Earmuffs, Leightning® Hi-Visibility L1HHV/L2FHV/L3HV
Supplier: Howard Leight by Honeywell
111-0396EA 32.7 EUR
DALL1013942 DALL1013941 DALL1015021
Earmuffs, Leightning® Hi-Visibility L1HHV/L2FHV/L3HV
In addition to noise exposure, there are other safety risks when working outside or with poor lighting conditions. Leightning Hi-Visibility earmuffs provide outstanding visibility day and night, whilst protecting your hearing.
  • Luminous green capsules provide particularly good visibility in darkness, poor light or poor weather conditions
  • Light, reflective headband offers an even higher level of safety, particularly in poor light
  • High level of comfort during extended wear; minimal pressure on the head
  • Robust steel headband for demanding requirements, particularly in rough conditions
  • Sealing cushion inserts are quick and easy to replace
  • Helmet-mounted earmuffs feature robust steel wire arms, able to withstand demanding use; can be used with a wide range of helmets
Air Flow Control™ technology ensures better, consistent damping for almost every industrial environment in which there is exposure to noise.
SNR: 28 dB (L1HHV)
SNR: 32 dB (L2HV)
SNR: 34 dB (L3HV)
Weight: 192 g (L1HVV), 236 g (L2HV); 304 g (L3HV)
Certifications: EN 352-1