Microplate readers, Luminoskan Ascent
Supplier: Thermo Scientific
LABH5300170EA 0 EUR
LABH5300170 LABH5300160 LABH5300172 LABH5300173
Microplate readers, Luminoskan Ascent
The Luminoskan Ascent is a robust, compact, microplate luminometer for luminometric research applications, such as reporter gene, immuno and cell based assays, enzyme studies, molecular interactions, nucleic acid quantification and microbiological assays.
  • Excellent optical performance
  • Versatile plate formats offered
  • Fast reading speeds
  • Up to three dispensers for reagent addition
  • Designed for robot compatibility
Ordering information: PC is not included with the instruments, but PC software (Thermo Scientific Ascent Software) is included.