You Searched For: Dissolved Oxygen Electrodes
33  results were found
SearchResultCount:"33"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Electrode, Dissolved Oxygen, VWR®, 4 m cable
Catalog Number: (664-0182)
Supplier: VWR Collection
Description: This IDP (Intelligent Digital Probe) dissolved oxygen/temperature sensor can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter mete...
UOM: 1 * 1 items
Catalog Number: (664-0042)
Supplier: VWR Collection
Description: Electrode, Dissolved Oxygen, VWR® PHenomenal®, pHenomenal OXY11-3, galvanic, 3 m cable, 8-pin
UOM: 1 * 1 items
Catalog Number: (664-0510)
Supplier: WTW
Description: Self-stirring oxygen probe for simultaneous stirring and measurement.
UOM: 1 * 1 items
Supplier: WTW
Description: Optical dissolved oxygen probes for field and laboratory applications. For use with IDS meters.
Supplier: WTW
Description: Galvanic dissolved oxygen sensors for universal (CellOx®) or field use (DurOx®).
Supplier: Mettler - Toledo
Description: Electrode, Dissolved Oxygen, LE621 (IP 67), galvanic, with NTC30 KΩ, 0 - 200%, 0 - 20 mg/l, 0...+50 °C, Ø×L: 12×120 mm, BNC/Cinch, 1 m cable
Supplier: HAMILTON BONADUZ
Description: These electrochemical oxygen sensors are ideal for applications with high demands for hygiene, e.g. in pharmaceutical industry, in biotechnology and i...
Supplier: Hach
Description: IntelliCALâ„¢ LDO101 is a digital, luminescent dissolved oxygen (LDO) electrode which is ideal for measuring dissolved oxygen in wastewater, drinking wa...
Supplier: Mettler - Toledo
Description: The InLab® OptiOx optical dissolved oxygen sensor is based on the reliable RDO® (Rugged Dissolved Oxygen) technology. RDO® is fast, precise and produc...
Catalog Number: (664-0205)
Supplier: Hach
Description: The LBOD101 is a digital, luminescent dissolved oxygen (LDO) sensor with a replaceable, integrated stirring system. The electrode measures the dissolv...
UOM: 1 * 1 items
Supplier: Mettler - Toledo
Description: The InLab® 605-ISM DO sensor incorporates the newest technology in polarographic dissolved oxygen determination.
Catalog Number: (HACHLZW5130970002)
Supplier: Hach
Description: Polarographic dissolved oxygen (DO) probe with built-in temperature sensor. For use with portable meters.
UOM: 1 * 1 items
Catalog Number: (SCOI285206410)
Supplier: SI Analytics
Description: Electrode, Dissolved Oxygen, Ox 1113T, −5...+45 °C, Ø×L: 12×120 mm, 1 m fixed cable with 8-pole plug
UOM: 1 * 1 items
Supplier: Endress+Hauser
Description: Memosens 2.0 hygienic sensor for the life sciences and food industries.
New Product
Catalog Number: (664-0153)
Supplier: Thermo Orion
Description: This BOD probe is ideal for accurately and conveniently conducting dissolved oxygen measurements. Self-stirring with One-Touch control, the highly eff...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 33
no targeter for Bottom