Anti-CEE Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BOSSBS-6405R-CY3EA 437 EUR
BOSSBS-6405R-CY3
Anti-CEE Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Get4 is a 327 amino acid cytoplasmic protein that exists as two alternatively spliced isoforms. Get4 forms a multiprotein complex, known as the BAT3 complex, with UBL4A, BAT3 and ARSA. The BAT3 complex plays a role in transporting tail-anchored membrane proteins to the endoplasmic reticulum membrane. The gene encoding Get4 maps to human chromosome 7p22.3. Human chromosome 7 houses over 1,000 genes, comprises nearly 5% of the human genome and has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CEE
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR