Anti-4-HNE Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BOSSBS-6313R-A647EA 436 EUR
BOSSBS-6313R-A647
Anti-4-HNE Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Aldehydic products of lipid peroxidation, such as 4 hydroxynonenal (4 HNE), have been implicated in the etiology of pathological changes under oxidative stress as a key mediator of oxidative stress induced cell death. It is a stable product of lipid peroxidation, is proarrhythmic and may contribute to the cytotoxic effects of oxidative stress

4-HNE has been hypothesized to play a key role in cell signal transduction, in a variety of pathways from cell cycle events to cellular adhesion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: 4-HNE
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR