Anti-WNT8A Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BOSSBS-6129R-CY5EA 437 EUR
BOSSBS-6129R-CY5
Anti-WNT8A Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Ligand for members of the frizzled family of seven transmembrane receptors. May play an important role in the development and differentiation of certain forebrain structures, notably the hippocampus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WNT8A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR