Anti-YWHAE Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BOSSBS-5590R-FITCEA 451 EUR
BOSSBS-5590R-FITC
Anti-YWHAE Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Adapter protein implicated in the regulation of a large spectrum of both general and specialized signaling pathway. Binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: YWHAE
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR