Anti-CD19 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BOSSBS-3065R-CY7EA 450 EUR
BOSSBS-3065R-CY7
Anti-CD19 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Assembles with the antigen receptor of B-lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD19
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR