SPE columns for special applications, J.T.Baker®, BAKERBOND spe™ PCB-A
Supplier: Avantor®
J.T.Baker®
7511-04EA 389.16 EUR
7511-04
SPE columns for special applications, J.T.Baker®, BAKERBOND spe™ PCB-A
Chromatography Columns
This solid phase extraction column is designed for the extraction of PCB’s in oil (dirty samples) in combination with SPE silica gel.