SPE columns for special applications, J.T.Baker®, BAKERBOND spe™ PAH Aqua
Supplier: Avantor®
J.T.Baker®
7490-07EA 286.64 EUR
7490-07 7490-08
SPE columns for special applications, J.T.Baker®, BAKERBOND spe™ PAH Aqua
Chromatography Columns
These solid phase extraction columns are designed for the extraction and clean-up of Poly Aromatic Hydrocarbons (PAH) from water, including the 16 priority pollutant EPA PAH’s and for several applications using different SPE columns.