J.T.Baker®, XE-Difference Controls
Supplier: Avantor
J.T.Baker®
3733.EA 36.6 EUR
3733. 3732. 3731.
J.T.Baker®, XE-Difference Controls
Clinical Diagnostic Supplies Clinical Reagents and Kits
XE-difference control is a tri-level whole blood control preparation intended for use in hemocytometry to monitor daily accuracy and precision of cell analysers and manual analytical procedures.
  • Composed of human RBC, mammalian WBC, and platelets
  • Cells are suspended in a plasma like fluid
  • Labeled with a barcode used for the automatic QC mode
Suitable for use on Sysmex XE 2100 XS 1000i and XT 1800/2000 analysers.
Delivery information: Supplied with assay values sheets.