Electrode storage solution, HI 80300L
Supplier: Hanna
620-3777EA 26.8 EUR
620-3777
Electrode storage solution, HI 80300L
Electrolytes Electrode Storage Solutions
This storage solution is supplied in an FDA compliant bottle. It is prepared with reagent grade chemicals that can be used to ensure optimum performance of pH and ORP electrodes.
  • Keeps the glass membrane of the pH electrode hydrated
  • Manufactured specifically for pH and ORP electrodes
  • Light blocking bottle to prevent against degradation by UV light
  • Air tight bottle with tamper-proof seal ensures quality and freshness of solution
  • Clearly marked expiration date and lot number
The storage solution is specifically formulated to minimise microbial growth and to prevent any effects of diffusion/osmotic from storing a probe in a solution with the highly concentrated inner reference electrolyte.