You Searched For: Hot Hand Protectors
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (112-1981)
Supplier: Saint-Gobain
Description: This heat resistant hand protector made from silicone rubber protects hands while handling hot labware.
UOM: 1 * 1 items
Catalog Number: (KART3915)
Supplier: KARTELL
Description: These hand protectors made of silicone rubber provide protection against hot containers up to +260 °C and cold containers down to –57 °C. Internal rel...
UOM: 1 * 1 items
Supplier: Bel-Art Products, a Part of SP
Description: This protector is made of durable silicone rubber which remains flexible from –57 to +260 °C.
Supplier: BÃœRKLE
Description: The cold and heat resistant HotGrip protectors are made of silicone rubber. They protect fingers and hands from burns and freezing when working with a...
Catalog Number: (AZLOPGR406)
Supplier: SciLabware
Description: Hand protector made of silicone rubber.
UOM: 1 * 1 items
Supplier: FUNAKOSHI & CO LTD
Description: These gripping mittens made from flexible silicone elastomer can be used to handle hot or cold objects and to carry them short distances. The shape on...
Catalog Number: (VWRU89026-680)
Supplier: VWR Collection
Description: This full-hand silicone rubber mitt is ideal for handling extremely hot or cold objects, from –57 to 260 °C, for brief periods of time. The mitt featu...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom