LEIC39601006
Supplier: Leica
LEIC39601006EA 66.8 EUR
LEIC39601006
LEIC39601006
Paraplast Regular is recommended for general tissue embedding. It is composed of a refined mixture of highly purified paraffin containing plastic polymers of regulated molecular weights. This supports improved tissue infiltration and superior quality sections with minimal compression.
Paraplast gives brittle, difficult to section tissue an innate elasticity, promoting wrinkle-free sections. It has a melting point of 56° C.