Anion exchange chromatography sorbents, HyperCel™ STAR AX
Supplier: Pall Laboratory
PALL20197-026EA 120 EUR
PALL20197-026 PALL20197-032
Anion exchange chromatography sorbents, HyperCel™ STAR AX
HyperCel™ STAR AX sorbent is designed for direct capture of proteins from bioreactor feed streams at moderate or higher salt conductivity.
  • High dynamic binding capacity (DBC) at short residence time (2 minutes or lower)
  • High flow rate properties for fast feedstock processing
Applications include direct capture of recombinant proteins, monoclonal and polyclonal antibodies, plasma derivatives or other biopharmaceuticals.