VWR®, PCR Plates, FrameStar
Supplier: Avantor
732-1125EA 217 EUR
VWRI732-1125
VWR®, PCR Plates, FrameStar
FrameStar PCR well plates are designed for high throughput PCR and are compatible with the majority of 384-well block PCR, qPCR and sequencing instruments. Designed to fit standard blocks, with PP wells, a skirted PC frame and a cut corner at A24.
  • Recommended for low volume PCR
  • Ultra-smooth, uniform, thin-walled PP tubes for optimum PCR and real-time (RT, qPCR) results
  • Compatible with standard multichannel pipettes
  • Improved seal integrity due to thermal stability of the frame
  • Rigid 2-component design eliminates warping and distortion during PCR making it ideal for use with robotic systems
Working volume: 30 µl.