Microbiological filtration capsules
514-0791EA 0 EUR
VWRI514-0791 VWRI514-0790 VWRI514-0789
Microbiological filtration capsules
These point-of-use tap and shower filters with sterilising grade PES membrane provide a secure method to minimise the risk of infection from waterborne bacteria such as Legionella and Pseudomonas and provide the highest protection to patients.
  • Indicated for 31 day use (62 day use for shower filter)