REDIchip™ (Resonance-Enhanced Desorption Ionisation) plates enables matrix-free small molecule analysis on a MALDI platform.
- Rapid data acquisition
- No matrix background
- Clean analyte peaks
- Definitive analysis
- Fits standard 25,4×76,2 mm microscope slide adapters
REDIchips™ are sample target plates with nanopost array (NAPA) surfaces for matrix-free laser desorption ionisation of small molecules. This robust alternative to sample analysis by traditional methods requires minimal sample preparation and is suitable for simple and complex mixtures. Using a matrix-free approach, small molecules can be rapidly and quantitatively analysed producing sensitive, reproducible results. It is easily integrated into existing MALDI-MS instrumentation using standard adaptors and the high-throughput, 96-target design features generous 2 mm spots for accuracy in sample spotting.