96-well plates, REDIchip™
Supplier: Protea Biosciences
PTEAREDI-1000-10EA 4823.88 EUR
PTEAREDI-1000-10 PTEAREDI-1000-1 PTEAREDI-1000-5
96-well plates, REDIchip™
Microplates Cell Culture Plates
REDIchip™ (Resonance-Enhanced Desorption Ionisation) plates enables matrix-free small molecule analysis on a MALDI platform.
  • Rapid data acquisition
  • No matrix background
  • Clean analyte peaks
  • Definitive analysis
  • Fits standard 25,4×76,2 mm microscope slide adapters
REDIchips™ are sample target plates with nanopost array (NAPA) surfaces for matrix-free laser desorption ionisation of small molecules. This robust alternative to sample analysis by traditional methods requires minimal sample preparation and is suitable for simple and complex mixtures. Using a matrix-free approach, small molecules can be rapidly and quantitatively analysed producing sensitive, reproducible results. It is easily integrated into existing MALDI-MS instrumentation using standard adaptors and the high-throughput, 96-target design features generous 2 mm spots for accuracy in sample spotting.