Filter photometers, photoLab®
Supplier: WTW
713-3002EA 2550 EUR
713-3002 713-3001
Filter photometers, photoLab®
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Filter photometer with six wavelengths for routine testing of wastewater and drinking water using cell tests.
  • Combines precision with high comfort and speeds
  • Easy to operate for occasional measurements
  • High test throughput at highest speeds
Can be used in water analytics as well as galvanization and in the food industry.
Certifications: AQA/IQC multi-level.