MALDI matrix, ultrapure sinapic acid
Supplier: Protea Biosciences
PTEACMS-101-1GEA 484.54 EUR
PTEACMS-101-1G CMS-101-5 PTEACMS-101-10
MALDI matrix, ultrapure sinapic acid
Protein Purification Protein Purification Reagents
Ultrapure Sinapic Acid is a recrystallised matrix for matrix assisted laser desorption ionisation (MALDI) mass spectrometry.
  • Improved MALDI–MS sensitivity (higher S/N)
  • Easy reconstitution directly in vial for ready to use stock
Applications include analysis of high molecular weight proteins.