MALDI matrix, ultrapure CHCA
Supplier: Protea Biosciences
PTEACMS-100-1GEA 484.54 EUR
PTEACMS-100-1G PTEACMS-100-5 PTEACMS-100-10
MALDI matrix, ultrapure CHCA
Protein Purification Protein Purification Reagents
Ultrapure α-cyano-4-hydroxycinnamic acid (CHCA) is a recrystallised matrix for matrix-assisted laser desorption ionisation (MALDI) mass spectrometry.
  • Improved MALDI–MS sensitivity (higher S/N)
  • Forms homogeneous crystals resulting in excellent resolution
  • Easy reconstitution directly in vial for ready to use stock
Research applications include MALDI mass spectrometry analysis of peptides and small proteins and post-source decay experiments.