Mobile phase sparge filters
Supplier: Restek
REST25312EA 40.9 EUR
REST25312
Mobile phase sparge filters
Filters Liquid Process Filters
The sparge filter is used to prepare and maintain mobile phases free of dissolved gas. Filters are made from 316 stainless steel and PEEK, and they are compatible with most solvents.
  • Connects to standard 3,2 mm (OD) PTFE tubing