Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Nucleic Acid Purification Systems
https://ru.vwr.com/stibo/smallweb/std.lang.all/95/97/18989597.jpg
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
Supplier:  GE Healthcare - Life Sciences
Description:   illustra™ PuReTaq Ready-To-Go™ PCR beads are designed for performing standard PCR. The only additional reagents are water, template DNA and primers. T...
Supplier:  GE Healthcare - Life Sciences
Description:   Sensitive, simple technique for the detection of genomic polymorphisms, using nanogram quantities of DNA.
Supplier:  GE Healthcare - Life Sciences
Description:   Ready-To-Go™ You-Prime First-Strand beads are preformulated, single-dose reaction beads prepackaged in thin-walled 0,5 ml tubes compatible with most t...
UOM:  1 * 50 Tests
Supplier:  G-Biosciences
Description:   The SpinOUT™ GT-100, GT-600 and GT-1200 columns are versatile, spin format columns for the desalting and buffer exchange of protein solutions ranging ...
Catalog Number: (USBM139281PK)
Supplier:  USB MOLECULAR BIOLOGY REAGENTS
Description:   With screw-on lid, Luer-lock lid, stopper, and filter insertion tools.
UOM:  1 * 1 PAC
Catalog Number: (EDVO625)
Supplier:  EDVOTEK EUROPE LTD
Description:   Each PCR Bead contains Taq DNA polymerase, Taq DNA polymerase buffer, dNTP mixture and MgCl₂. Pack contains sufficient for 25 reactions.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6  of 6