You Searched For: Rotors
150  results were found
SearchResultCount:"150"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: VWR®, Swing rotor for microplates with carriers, max. RCF 2200 ×g, max. rpm 5000 max. capacity 4 microtitre or 2 deep well plates (max. height: 31,5 mm)
Supplier: VWR Collection
Description: VWR®, Rotor for 4× (8×0,2 ml) PCR strips, max. 6000 RPM, For: MicroStar 12
Supplier: VWR Collection
Description: Replacement tube rotor, quick release, 8×1,5/2,0 ml tubes, for MiniStar blueline centrifuge
Supplier: VWR Collection
Description: VWR® MiniStar silverline, Rotor for 4 strips of 8×0,2 ml tubes, For: MiniStar silverline
Supplier: Thermo Scientific
Description: General purpose centrifuge rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapter...
Sale
Catalog Number: (521-4070)
Supplier: Beckman Coulter
Description: Optimaâ„¢, SW 32 Ti swinging bucket rotor assembly. Includes: SW 32 Ti rotor bucket set, For: Ultracentrifugation
UOM: 1 * 1 items
Supplier: Beckman Coulter
Description: JA 12 fixed angle (high performance) rotor with Biosafety lid 12×50 ml for use in Avanti J Series and J2 Series centrifuges
Catalog Number: (521-4165)
Supplier: Beckman Coulter
Description: JS-5.3 swinging bucket (high performance) aluminium rotor package with microplate kit. For use in Avanti J-E and J-26 XP Series centrifuges only. Package includes rotor, buckets, tie-down knob, and microplate carrier kit (368914)
UOM: 1 * 1 items
Supplier: SORVALL
Description: Swinging bucket rotor, 8×2000 ml, 3900 min-1, 5374 ×g, incl. oval buckets and 2000 ml bottles, For: Sorvall™ BIOS 16
Supplier: THARMAC GmbH
Description: Closed, removable 12-position rotor needs adaptor (512-0727)
Catalog Number: (521-3987)
Supplier: Beckman Coulter
Description: JS-24.15 swinging bucket (high performance) aluminium rotor, titanium buckets. For use in Avanti J Series centrifuges (except Avanti J-E and Avanti J-HC). Reaches 24000 rpm in the Avanti J-30I centrifuge only. Not recommended for use in Avanti J-25 Series (max speed 10000 rpm), or Avanti J-26 XP Series (max. speed 10000 rpm). Incompatible with all J2 Series
UOM: 1 * 1 items
Catalog Number: (521-4078)
Supplier: Beckman Coulter
Description: Optimaâ„¢, SW 32 Ti rotor package, For: Ultracentrifugation
UOM: 1 * 1 items
Supplier: Beckman Coulter
Description: J-LITE JLA-16.250 fixed angle (high performance) aluminium rotor, with single lock lid, 6×250 ml, 16000 rpm, 38400 ×g. For use in Avanti J Series and J2 Series centrifuges. Maximum speed is 14000 rpm in J2 Series centrifuges
Catalog Number: (521-3805)
Supplier: Beckman Coulter
Description: JS-13.1 swinging bucket (high performance) aluminium rotor with carrying handle. For use in Avanti J Series and J2 Series centrifuges
UOM: 1 * 1 items
Catalog Number: (521-3991)
Supplier: Beckman Coulter
Description: Optima™, VTi 65.2 vertical tube rotor package, 65000 rpm, 416000 ×g, 16×5,1 ml. For use in Optima series ultracentrifuges and/or instruments classified: HRS, For: Ultracentrifugation
UOM: 1 * 1 items
Catalog Number: (521-4041)
Supplier: Beckman Coulter
Description: Major applications: Large volume differential sedimentation (pelleting) of subcellular organelles and viruses.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 150
no targeter for Bottom