Hazardous substance collection stations, HazMat

Supplier: CEMO
CEMG7597EA 1760 EUR
CEMG7597
Hazardous substance collection stations, HazMat
These Hazardous substance collection stations are approved for storage of all waterpolluting as well as flammable substances (previously classed as AI, AII, AIII and B).

  • Made of robust, lockable EW2-GH/VbF painted sheet steel construction
  • Variable internal equipment provides a universal solution
  • Ground clearance (100 mm) for safe transport
  • With containment volume for storing a maximum two 225 l barrels

W×D×H (ext.): 1420×890×1510 mm

W×D×H (int.): 1380×850×1070 mm
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR