Mycoplasma Stain kit
ICNA093030000EA
914
EUR
ICNA093030000
Mycoplasma Stain kit
Mycoplasma Stain kit
Mycoplasma stain is a DNA fluorochrome designed for in situ detection of mycoplasma and other prokaryotic organisms in cell cultures.
Highly specific for DNA as it occupies one of the grooves of the DNA double helix.
Ordering information: Includes a Hoehst stain 10 ml (0,5 µg/ml), 10X HBSS (10 ml), mounting medium (10 ml) and control slide.