Abbe refractometers, 60 series
Supplier: Bellingham + Stanley
635-0394EA 0 EUR
BELL10-99-61-191 BELL10-03-61-193 BELL10-99-61-193 BELL10-03-61-191
Abbe refractometers, 60 series
Abbe refractometers 60/DR and 60/95 have applications in quality control or research for the measurement of refractive indices in sugar solutions and liquid food, together with a wide range of chemicals.
Externally mounted LED source with compensation device to ‘achromatise' to the sodium wavelength (589 nm).
Delivery information: Supplied with external power supply, digital thermometer, traceable standardisation plate, contact liquid and instructions.