Pocket thermometers
Supplier: Hach
HACH189500EA 29.2 EUR
HACH189500
Pocket thermometers
Glass thermometers with permanently fused markings. Red or blue spirit-filled (kerosene and non-toxic dye).
Delivery information: Supplied in a heavy duty case.