Analogue stopwatch, Kienzle® E-8
LIMME-8EA 390 EUR
LIMME-8
Analogue stopwatch, Kienzle® E-8
This Kienzle® table stopwatch is a robust watch with shatterproof glass display and robust aluminium casing.
  • Black easy to read display
  • Clearly visible hands: Red second hand and yellow minute hand
  • Measuring range: 1 second to 60 minutes
This stopwatch functions with the aid of a mechanical, manual winding movement using a key on the back. Runs for 24 hours without using batteries.
H×W: 220×209,5 mm