Syringe filters
Supplier: Corning
516-1961EA 158.38 EUR
516-1961 516-1956 516-1955 516-1951 516-1950 516-1958 516-1957 516-1952 516-1953 516-1959 516-1954 516-1960
Syringe filters
Filters Syringe Filters
Syringe filters are available with a choice of membrane type: Polyethersulfone (PES) for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA), with or without prefilter, for lowest protein binding; Polytetrafluoroethylene (PTFE) for chemical resistance; regenerated cellulose (RC) – the best choice for DMSO compatibility; and nylon (NY) which is hydrophilic, surfactant-free and lowest extractable.
  • 100% integrity tested
  • Certified non pyrogenic and non cytotoxic
Manufactured in accordance with ISO 9002 standards
516-1959 is recommended as in-line air filter.