You Searched For: pH Electrodes
147  results were found
SearchResultCount:"147"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: These IDP (Intelligent Digital Probe) pH/temperature electrodes can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.
Supplier: VWR Collection
Description: Laboratory and field electrodes for general applications. Supplied with a fixed cable or plug head, VWR® electrodes can be connected to most pH meters...
Supplier: VWR Collection
Description: With 1 m fixed cable.
Supplier: VWR Collection
Description: pH electrodes with 1 m fixed cable or S7 connector.
Supplier: VWR Collection
Description: Combined pH electrodes with 1 m fixed cable.
Supplier: VWR Collection
Description: pH electrodes with BNC connector and cable.
Supplier: VWR Collection
Description: These pH electrodes are available with refillable electrolyte or non refillable.
Supplier: VWR Collection
Description: Combined electrodes providing fast, accurate and stable pH measurements, ideal for control quality and research and development use.
Catalog Number: (662-1381)
Supplier: VWR Collection
Description: This flat surface electrode is a combination pH/reference electrode with double reference junction design. It includes both a pH and reference half ce...
UOM: 1 * 1 items
Catalog Number: (662-1796)
Supplier: VWR Collection
Description: Electrode, pH, VWR®, Double junction reference electrode, half cell, 1 m cable, Glass, Electrolyte: 3,5 mol/l KCl, Ø×L: 12×150 mm, 3 mm pin, For: Water, creams, beverage, TRIS buffer
UOM: 1 * 1 items
Supplier: WTW
Description: These proven and tested pH electrodes are easy to use and offer high quality and precision.
Supplier: WTW
Description: High quality, enhanced sensor technology combined with most modern measurement electronics. The electrodes feature a plug head which can either be con...
Catalog Number: (662-1268)
Supplier: WTW
Description: Electrode, pH, TA-SensoLyt® SEA, - °C, -, Electrolyte: Polymer, -, For: Installation in former depth armature series TA 197-pH
UOM: 1 * 1 items
Supplier: WTW
Description: High quality, enhanced sensor technology combined with most modern measurement electronics.
Supplier: WTW
Description: These special pH electrodes extend the range of the IDS series for enhanced applications in Life Sciences and related applications.
Supplier: SI Analytics
Description: Autoclavable high-tech electrodes with outstanding electrical and physical performance characteristics guaranteeing exceptional accuracy and reliabili...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 147
no targeter for Bottom