You Searched For: ISE Electrodes
49  results were found
SearchResultCount:"49"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: WTW
Description: These easy-to-use, robust, high quality ion-selective electrodes are ideal for field applications.
Catalog Number: (662-0381)
Supplier: WTW
Description: These 500 series half cells require a separate reference electrode type R 503 (an exception for this is the gas-sensitive NH 500/2 with integrated ref...
UOM: 1 * 1 items
Catalog Number: (662-1595)
Supplier: Thermo Orion
Description: These electrodes are designed for measuring total residual chlorine quickly and accurately.
UOM: 1 * 1 items
Supplier: Thermo Orion
Description: These electrodes are designed for measuring cupric ions in aqueous solutions quickly, simply, accurately and economically.
Supplier: Thermo Orion
Description: These calcium electrodes are designed for measuring calcium in aqueous solutions simply, accurately and economically.
Catalog Number: (ORIO9305BN)
Supplier: Thermo Orion
Description: This fluoroborate half-cell can be easily and economically rejuvenated.
UOM: 1 * 1 items
Supplier: Mettler - Toledo
Description: These ion-selective half-cells (exception: Sodium-sensitive electrode) consist of two elements: a universal shaft and an ion-specific membrane module.
Supplier: SI Analytics
Description: Electrode, ISE, Ca 1100 PLH, Parameter (ISE): Calcium, pH: 2,5 - 11, 0...+40 °C, Plug head, For: -
Supplier: Hach
Description: IntelliCALâ„¢ ISEF121 is a digital combination fluoride ion selective electrode (ISE) with a non-refillable, single junction reference and built-in temp...
Supplier: Mettler - Toledo
Description: These ion selective half cells with solid-state membrane allow a quick and easy determination of concentration and activity of a variety of dissolved ...
Supplier: Hach
Description: IntelliCALâ„¢ ISECL181 is a digital, combination chloride ion selective electrode (ISE) with a non refillable, double junction reference and built-in te...
Supplier: Mettler - Toledo
Description: Combined ion-selective electrodes consisting of a measuring electrode complete with an integrated reference.
Supplier: Thermo Orion
Description: ISE half-cell electrodes for use with analysers.
Catalog Number: (HACHE41M003)
Supplier: Hach
Description: This ion selective electrode (ISE) is a half-cell with plastic body, a non-refillable internal electrolyte, and a chloride ion (Cl-) specific solid-st...
UOM: 1 * 1 items
Supplier: Thermo Orion
Description: These electrodes are designed for measuring lead ions in aqueous solutions quickly, simply, accurately and economically.
Catalog Number: (662-1591)
Supplier: Thermo Orion
Description: This electrode measures carbon dioxide, carbonate and bicarbonate in aqueous solutions quickly and accurately.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 49
no targeter for Bottom