Solid black and white microplates, 96-well
Supplier: Corning
Corning® NBS™ Corning®
CORN4591EA 600.67 EUR
CORN4591 734-1662 734-1663 CORN3789A 732-5522 734-1666 732-5517 734-1667 732-5596 734-4169 734-1643 732-5515
Solid black and white microplates, 96-well
Microplates Immunology Microplates and Strips
PS, without lid, non sterile. Available either with NBS™ non binding surface, ideal for reducing protein and nucleic acid binding at low concentrations, and for increasing assay signal-to-noise ratio, or with High Bind surface for immunological applications. Also available untreated for solution based assays.
Available either with NBS™ non binding surface, ideal for reducing protein and nucleic acid binding at low concentrations, and for increasing assay signal-to-noise ratio, or with High Bind surface for immunological applications. Also available untreated for solution based assays.