Storage microplates, 96-well, Corning®
Supplier: Corning
Corning®
734-1516EA 232.67 EUR
734-1516 734-1518
Storage microplates, 96-well, Corning®
Microplates Deep Well Microplates
PVC, clear, non sterile, without lid. Untreated microplates for solution-based assays, serial dilutions, and general storage applications.
Well volume: 250 µl (260 μl for V-bottom)
Recommended working volume: 50 to 150 µl