Microplates, 96-well, half area
Supplier: Corning
Corning® NBS™ Corning®
734-4159EA 1203.81 EUR
734-4159 734-4155 734-1622 734-4171 734-4154 734-1629 734-1626 734-4156 734-1628 734-1623 734-4157 734-4158 734-1625 734-1627 734-4117 734-4153 734-4170 734-4160 734-1624 734-4172
Microplates, 96-well, half area
Microplates HTS Microplates
PS, flat bottom, low volume microplates available with solid or clear bottom.
Recommended working volume: 25 to 125 µl