Cell culture plates, 96-well, for fluorescent and luminescent applications
Supplier: Corning
Corning®
734-1665EA 657.38 EUR
734-1665 734-1660 734-1609 734-1610 734-1549 734-1664 734-1661
Cell culture plates, 96-well, for fluorescent and luminescent applications
Microplates Cell Culture Plates Standard Plates
PS.
  • Black plates are designed to lower background in fluorescent assays
  • White plates are designed for luminescent assays
  • Treated for optimal cell attachment
  • Sterilised by gamma irradiation
  • Certified non pyrogenic