You Searched For: HTS Microplates
83  results were found
SearchResultCount:"83"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Manufactured from high quality crystal polystyrene. They are ideal for routine assays, mixing and storage.
Sale
Supplier: Avantor
Description: These plates are manufactured from high purity PP for efficient sample preparation and are suitable for use with magnetic particle processors such as ...
Supplier: VWR Collection
Description: PP, non coated, non sterile, chromatography tested and certified. Manufactured from ultra low bleed high purity basis resin and fully lot tested by GC...
Supplier: Greiner Bio-One
Description: Cycloolefin, black pigmented frame, 190 μm ultra-clear film bottom, tissue culture treated, sterile. SCREENSTAR microplates are specialised products f...
Catalog Number: (GREI792870-906)
Supplier: Greiner Bio-One
Description: Clear, 1536-well cycloolefin storage plates (with optimised geometry for low evaporation) for acoustic liquid handling.
UOM: 1 * 60 items
Supplier: Enzo Life Sciences
Description: Screen-Wellâ„¢ REDOX library contains 84 compounds with defined prooxidant or antioxidant activity.
Supplier: Enzo Life Sciences
Description: SCREEN-WELL® Endocannabinoid Library contains 58 compounds with defined, putative, potential or speculative activity at cannabinoid (CB) and vanilloid...
Supplier: Greiner Bio-One
Description: PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, ...
Supplier: Greiner Bio-One
Description: Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity b...
Supplier: Molecular Devices
Description: SpectraDropâ„¢ micro-volume microplate consists of aspecially designed adapter, PTFE-coated bottom slide, and top slide with evaporation-reducing spacer...
Catalog Number: (734-0984)
Supplier: Corning
Description: HTS microplate, Square well, Falcon®, No. of wells: 96, Well volume: 120 µl, Flat bottom, with lid
UOM: 1 * 5 items
Supplier: KARTELL
Description: Microtitre plate, Individually wrapped, Sterile, No. of wells: 96, PS, V-bottom, Clear
Supplier: Greiner Bio-One
Description: Characterised by low protein, DNA, RNA, and peptide binding properties, the non binding microplate surface significantly increases assay sensitivity b...
Supplier: Greiner Bio-One
Description: Optically clear borosilicate glass bottom (light path 175±15 µm) and black PS frame.
Supplier: Greiner Bio-One
Description: PS. With the exception of the Small Volumeâ„¢, all 384-well PS microplates have rounded square wells, which combine the advantages of flexible working v...
Sale
Supplier: Greiner Bio-One
Description: PS, without lid. Ideal for high throughput screening applications, these plates are available as clear bottom variants (in clear PS) and black or whit...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 83
no targeter for Bottom