VWR® Sample Splitters
Supplier: Avantor
HAVE205322303EA 692 EUR
HAVE205322303 HAVE205322334 HAVE205322327 510-0494 510-0487 510-0488 HAVE205322280 510-0489 HAVE205322273 510-0490 510-0491 HAVE205322310 510-0492 HAVE205322297 510-0493 HAVE205322341
VWR® Sample Splitters
Sample Dividers
For dividing sieved material into representative test sieve samples.
Type RT 6,3 and 12,5: Consists of 1 lacquered sheet steel rack, 300×250×270 mm, 1 stainless steel divider and 3 receivers which are tin sheet (1,75 litres) or laminated plastic (2 litres).
Type RT 25; 37,5; 50 and 75: Consists of 1 rack 620×260×420 mm, 1 divider and 3 receivers with grab handles (8 litres), all of galvanised steel.