Labelling tape cartridge, VERSAPRINTER™ & LABELIZER® plus
Supplier: Brady
BRDY142771EA 280.79 EUR
BRDY142771 BRDY142770 BRDY142758 BRDY142768 BRDY142757 BRDY142766 BRDY142774
Labelling tape cartridge, VERSAPRINTER™ & LABELIZER® plus
These tapes can survive the harshest environmental conditions indoors or outdoors. These tapes come in an array of materials, sizes and colors to work with multiple applications.
  • Temperatures from 82 to –40 °C
  • Conforms well to irregular, curved or rough surfaces
  • Adheres to pipes, walls, equipment, windows etc
It is ideal for general purpose industrial uses and stands up to grease, oil and most industrial chemicals.
Length: 27,0 m
Different colours available.