Solid phase extraction cartridges, activated sodium sulfate
Supplier: Restek
REST26207EA 0 EUR
REST26207
Solid phase extraction cartridges, activated sodium sulfate
Chromatography Columns
This closed end SPE cartridge is prepacked with approximately 2 grams of activated sodium sulphate in a convenient capped cartridge with both male and female Luer ends for easy connection to a variety of devices or equipment. The adsorbent is fully activated and ready to use for removal of excess water from organic solvent solutions, prior to many types of analysis.