The TQC automatic cupping tester allows to perform a cupping (Erichsen / Dent) test on coated steel panels to define the resistance of paint, varnish or related products to cracking and/or detachment from a metal substrate when subjected to gradual deformation by indentation under standard conditions.
- Driven by a micro-step controlled electro motor which allows precise and steady deformation with 0,01 mm steps
- Operation is intuitive by means of a jog-dial switch and a multi-lingual operating menu on a large illuminated display
- Easy-to-use with possible user calibration
- Manual and preset mode with integrated multi colour LED lighting
- Multiple languages
- Panel holder adjustable in angle
- Mandatory test in Qualicoat and QIB accredited laboratories
The test is either used as a “pass / fail” test by testing to a specified depth or defining the minimum depth at which a coating fails by gradually increasing the indentation. The ISO 1520 standard requires panels to be slowly deformed at a steady rate between 0,1 mm/s and 0,3 mm/s without interruption. Especially with thicker steel panels hand-operated testers not always allow an uninterrupted deformation.
An integrated LED powered sample illumination system comforts examining the coating under test. To guarantee maximum visibility of all possible types of surface including high gloss, matte or colored samples the angle of the LED light can be set. Choose light from just one or all directions. The strength of the LED’s is adjustable but also the colours can be changed to achieve maximum contrast.
Certifications: Tester includes standards such as EN-ISO 1520, DIN 53156, DIN 53232, BS 3900-E4, NBN T22-104, NFT 30 019.
Delivery information: Tester is supplied with reference panel, optical tool support rod, optical tool mount, power cord, manual and calibration certificate.