Label IT® µArray Labeling Kits, Cy®3 and/or Cy®5
MIRUMIR8750EA 0 EUR
MIRUMIR8750 MIRUMIR8850 MIRUMIR8710 MIRUMIR8225 MIRUMIR8205 MIRUMIR8810
Label IT® µArray Labeling Kits, Cy®3 and/or Cy®5
Label IT® µArray is an optimised labeling kit designed for dual-channel microarray applications using Cy®3 and/or Cy®5 dyes.
  • Easily and consistently achieve optimized labeling densities for microarray applications
  • Confidently detect rare transcripts and small changes in gene expression
  • Suitable for labeling mRNA, cDNA and cRNA templates with Cy®3 and Cy®5 dyes for expression profiling analysis
Delivery information: Each kit contains Label IT® Cy®3 and/or Cy®5 reagents, reconstitution solution, 10X labeling buffer M, reagent D1, buffer N1, 0,5M EDTA, 10X STOP reagent and 5X fragmentation buffer.