Label IT® µArray Biotin Labelling Kit
MIRUMIR8050EA 0 EUR
MIRUMIR8050 MIRUMIR8010
Label IT® µArray Biotin Labelling Kit
Label IT® µArray Biotin Labelling Kit is optimised for single-channel microarray applications using biotin. The kit is suitable for labelling mRNA, cDNA and cRNA templates that are used for single-channel expression profiling analyses.
  • Easily and consistently achieve optimal labeling densities for microarray applications
  • Confidently detect rare transcripts and small changes in gene expression
Delivery information: Each kit contains Label IT® µArray Biotin Reagent, Reconstitution Solution, 10X Labeling Buffer M, Reagent D1, Buffer N1, 0.5M EDTA and 5X Fragmentation Buffer. Biotin detection reagents are not included.