Label IT® µArray Dual Labeling Kit
MIRUMIR8125EA 0 EUR
MIRUMIR8125 MIRUMIR8105
Label IT® µArray Dual Labeling Kit
Label IT® µArray Dual Labeling Kit is optimised for dual-channel microarray applications using biotin and fluorescein. The kit is suitable for labeling mRNA, cDNA and cRNA templates with biotin and fluorescein dyes for expression profiling analysis.
  • Easily and consistently achieve optimal labeling densities for microarray applications
  • Confidently detect rare transcripts and small changes in gene expression
Delivery information: Each kit contains Label IT® Biotin and Fluorescein Reagents, Reconstitution Solution, 10X Labeling Buffer M, Reagent D1, Buffer N1, 0.5M EDTA, 10X STOP Reagent and 5X Fragmentation Buffer. Biotin and Fluorescein detection reagents are not included.