Cleanroom frocks, Tyvek® IsoClean®, model IC 270 B
Supplier: DuPont
DUPOIC270BWH00/LEA 235 EUR
DUPOIC270BWH00/L DUPOIC270BWH00/XL DUPOIC270BWH00/2X DUPOIC270BWH00/3X DUPOIC270BWH00/S DUPOIC270BWH00/M
Cleanroom frocks, Tyvek® IsoClean®, model IC 270 B
These single-use frocks deliver an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non hazardous light liquid splash.
  • Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
  • Elastic openings for tighter fit at wrist
  • Bound neck for lower particle shedding
  • Adjustable snap at neck for better fit
  • Body-length jacket design with front snap closures for easy donning and doffing
Suitable for use in ISO class 6 - 9, GMP class C/D cleanroom environments.
Length: 99 to 114 cm
Weight: 45 g/m²
Certifications: (EU) 2016/425; partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents)
Packaging: Bulk packaged in double transparent poly liners. 30 per case.