These single-use frocks deliver an ideal balance of protection, durability and comfort. Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non hazardous light liquid splash.
- Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration
- Elastic openings for tighter fit at wrist
- Bound neck for lower particle shedding
- Adjustable snap at neck for better fit
- Body-length jacket design with front snap closures for easy donning and doffing
Suitable for use in ISO class 6 - 9, GMP class C/D cleanroom environments.
Length: 99 to 114 cm
Weight: 45 g/m²
Certifications: (EU) 2016/425; partial body chemical protective clothing, Category III, Type PB [6-B]; EN 14126 (barrier to infective agents)
Packaging: Bulk packaged in double transparent poly liners. 30 per case.