VWR®, dNTP set
Supplier: Avantor
VWR®
K461-KITEA 213 EUR
K461-KIT
VWR®, dNTP set
Nucleic Acid Reagents Nucleotides
Set contains one vial of each dNTP (dATP, dTTP, dCTP, dGTP), 40 μmol of 100 mM solution, pH 7,0.
  • Building blocks for the amplification of DNA
  • High purity grade
DNA consists of a sugar molecule (deoxyribose), a nitrogenous base (purine in adenine and guanine, pyrimidine in thymine or cytosine) and a phosphate molecule.
Delivery information: Kit includes four separate tubes containing the following:
1×dATP, 40 µmol of 100 mM Solution, 400 µl, pH 7,0
1×dTTP, 40 µmol of 100 mM Solution, 400 µl, pH 7,0
1×dCTP, 40 µmol of 100 mM Solution, 400 µl, pH 7,0
1×dGTP, 40 µmol of 100 mM Solution, 400 µl, pH 7,0