Surface roughness comparator
Supplier: Industrial Physics
SIMELD6009EA 265 EUR
SIMELD6009
Surface roughness comparator
Comparison standard for surface roughness comparator consisting of six grit- and shot-blasting.
  • High nickel test plate
  • Grit and short range scales
  • Leather pouch to protect the standard when not in use
Certifications: NF/E05-051 ; ISO 2632 / I 1975 and 2632/II 1977
Delivery information: Supplied with table and manual in a leather pouch.