Disposable particle filtering respirators, 9320
Supplier: 3M
111-0034EA 69.7 EUR
MMMA9320
Disposable particle filtering respirators, 9320
The disposable, foldable FFP2 respirator protects against dusts, mists, smoke and fume. It can be used in certain applications against some bio-aerosols such as the influenza virus.
  • Unique 3-panel design fits a wide range of face shapes and sizes, accommodates facial movements and makes the respirator easy to talk through
  • Concealed adjustable nose-clip is metal detectable
  • Coloured straps for easy identification of protection class (blue for P2)
  • Made from 3M™ Advanced Electret Filter Material for effective filtration with low breathing resistance
  • Does not contain components made from natural rubber latex
Protection factor: 10 x Workplace Exposure Standard
Certifications: AS/NZS 1716:2012