Bis-TRIS hydrochloride ≥98.0%, Biotechnology Grade

N108-100GEA 140 EUR
N108-100G
Bis-TRIS hydrochloride ≥98.0%, Biotechnology Grade
Bis-TRIS hydrochloride
Formula: C₈H₂₀NO₅Cl
MW: 245,7 g/mol
MDL Number: MFCD00171252
CAS Number: 124763-51-5

Order Now

Specification Test Results

Abs.@260nm (0,1M, Water) ≤0,05 AU
Abs.@290nm (0,1M, Water) ≤0,03 AU
pH (1.0%, Water) @25°C 3,8 - 4,4
Purity ≥98 %
Solubility (0.1M, Water) Pass
Water Content (KF) ≤1,0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR