Silicone-free laboratory grease, glisseal HV
Supplier: Borer
291-1112EA 316 EUR
291-1112 DECO514310.00-CA15 DECO514310.00-CA1U
Silicone-free laboratory grease, glisseal HV
Lubricants
Silicone-free laboratory grease for sealing and lubricating fixed and rotary ground glass joints and for greasing fittings made of plastics and metall. Withstands high-vacuum conditions.
  • Apply only a very thin layer
  • Withstands high-vacuum conditions, specifically suitable for applications under high vacuum
  • Excellent thermal stability, constant lubricating consistency between -40 °C and +220 °C, in the short run even up to 320 °C
  • No dripping point
  • Chemically stable against most acids, alklalis and gases
  • Silicone-free
  • Can be easily removed with the deconex® cleaning concentrates
  • Technical support from the manufacturer and distributor